Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (6)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
surgical-masks
https://si.vwr.com/stibo/smallweb/std.lang.all/95/95/18709595.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"12"
  List View Grid View Easy View (new)
Sort by:
Catalog Number: (111-8940)
Supplier:  3M MEDICAL
Description:   The efficient filtering and good facial fit of these masks give patients optimal protection and maximum comfort.
UOM:  1 * 50 items
Catalog Number: (111-9004)
Supplier:  KIMBERLY CLARK
Description:   Designed with extra lightweight materials to enhance breathability, this non-sterile surgical mask provides a cooler wearing comfort.
UOM:  1 * 50 items
Supplier:  Nitritex
Description:   Profile™ PFM surgical face mask with highly efficient bacterial filtration.
Catalog Number: (ANSM85005)
Supplier:  ANSELL MEDICAL PRODUCTS
Description:   These speciality masks are designed to prevent transmission of disease from a health care worker to a patient. (e.g. spit, mucous).
UOM:  1 * 50 items
Supplier:  Zellstoff-Vertriebs-GmbH & Co. KG
Description:   Made from non-woven, spunbound PP, they are comfortable to wear, safe and highly efficient.
Supplier:  ANSELL MEDICAL PRODUCTS
Description:   Splash and penetration resistant care masks. Inner surface made from 40% cellulose and 60% polyester; outer surface made from PP. PP filter contains n...
Catalog Number: (ANSM00152)
Supplier:  ANSELL MEDICAL PRODUCTS
Description:   These polyester standard cone masks with headbands and soft flared edges provide a comfortable fit.
UOM:  1 * 50 items
Supplier:  ANSELL MEDICAL PRODUCTS
Description:   The masks feature a flexible nosepiece for effective sealing and polyester cellulose inner layer to help prevent irritation.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 12
  1  2  Next