Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (18)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "polymerases"
https://si.vwr.com/stibo/smallweb/std.lang.all/96/65/18989665.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"40"
  List View Grid View Easy View (new)
Sort by:
Supplier:  VWR Chemicals
Description:   VWR® Taq DNA Polymerase is an ultra-pure, thermostable, recombinant DNA polymerase, which provides robust PCR performance in a wide range of PC...
Supplier:  VWR Chemicals
Description:   AccuPOL DNA Polymerase is a thermostable enzyme that possesses 3' - 5' exonuclease proofreading ability, which enables the polymerase to correct nucle...
Supplier:  VWR Chemicals
Description:   VWR® TEMPase Hot Start DNA Polymerases are highly stable Polymerases, featuring higher specificity, superior sensitivity and greater yields compared t...
Supplier:  VWR Peqlab
Description:   peqGOLD Pure Taq DNA polymerase is a highly purified version of peqGOLD Taq, certified completely free of contaminating E. coli D...
Supplier:  VWR Chemicals
Description:   Taq DNA polymerase is a thermostable enzyme that can withstand prolonged incubation at temperatures up to 95°C without significant loss of acti...
Catalog Number: (PEQL04-1090)
Supplier:  VWR Peqlab
Description:   peqGOLD Uracil DNA-Glycosylase (UDG) in combination with dUTP prevents contamination in PCR reactions.
UOM:  1 * 1 items
Catalog Number: (732-3262)
Supplier:  VWR Peqlab
Description:   Originally isolated from the thermophilic archaebacteria Pyrococcus woesei, peqGOLD Pwo DNA Polymerase exhibits high levels of 3‘→5‘ exo...
UOM:  1 * 1 items
Supplier:  VWR Peqlab
Description:   A blend of peqGOLD 'Standard' Taq and an inert red dye enables quick PCR set-up and avoids the need for additional dyes for electrophoresis.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 40
  1  2  3  4  5  Next