Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "plasmid-purification-kits"
https://si.vwr.com/stibo/smallweb/std.lang.all/96/55/18989655.jpg
VWR’s huge choice of plasmid purification kits range from well-established miniprep spin-column kits to midi or maxi prep kits.
Miniprep kits will provide sufficient yield and purity for most applications in molecular biology, such as cloning or sequencing. Midi or maxi preps (also available in endotoxin-free versions) will result in higher yields of intact plasmid, fulfilling the requirements for transfecting sensitive cells and long-term storage.
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"36"
  List View Grid View Easy View (new)
Sort by:
Supplier:  VWR Chemicals
Description:   The E-Z 96® S.E. Plasmid DNA Isolation Kit is designed for rapid high-throughput purification of plasmid DNA from small volume bacterial cultures. The...
UOM:  1 * 1 KIT
Catalog Number: (D6947-01)
Supplier:  VWR Chemicals
Description:   The E.Z.N.A.® X-Press Plasmid Kit uses a novel method for very fast and easy plasmid preparation. The X-Press plasmid isolation method replaces the tr...
UOM:  1 * 50 Tests
Supplier:  VWR Chemicals
Description:   This second generation plasmid miniprep kit yields 40 - 70 µg of high-quality DNA in less than 30 minutes using the same format as the E.Z.N.A.® Plasm...
Supplier:  VWR Chemicals
Description:   The E.Z.N.A.® BAC/PAC DNA Kit is designed for rapid high throughput purification of BACs, PACs, and P1s from small volume bacterial cultures. This kit...
Supplier:  VWR Chemicals
Description:   The E.Z.N.A.® HP Plasmid Mini Kit I is designed to isolate up to 30 µg of high-quality plasmid DNA from 1 to 5 ml bacterial cultures from strains...
UOM:  1 * 50 Tests
Supplier:  VWR Chemicals
Description:   E.Z.N.A.® Fastfilter® Plasmid kits rapidly purify plasmid DNA utilising lysate clearance filter syringes, whilst E-Z 96® Fastfilter Plasmid kits emplo...
Supplier:  VWR Peqlab
Description:   XChange Plasmid Maxi Kit for the isolation of up to 520 µg plasmid DNA per preparation. Anion exchange technology ensures highly pure and intact plasm...
Supplier:  VWR Chemicals
Description:   Plasmid isolated with traditional purification procedures normally contain high levels of endotoxins (also known as lipopolysaccharides or LPS) that c...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 36
  1  2  3  4  5  Next