You Searched For: Sanger Sequencing Reagents
6  results were found
SearchResultCount:"6"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (MAG-FRAG-I-5)
Supplier: Corning
Description: The AxyPrepâ„¢ FragmentSelect Kits utilise a paramagnetic bead technology for quick high throughput optimised DNA size selection suitable for various ne...
UOM: 1 * 1 KIT
Catalog Number: (25-9005-98)
Supplier: Cytiva
Description: AutoScreen 96 well plate consists of a 96-well filter plate containing DNA grade Sephadexâ„¢ G-50 for purification of sequencing reactions prior to anal...
UOM: 1 * 10 items
Catalog Number: (786-14)
Supplier: G-Biosciences
Description: SG-Lysine-Câ„¢ endopeptidase, from Lysobacter enzymogenes, is a serine protease highly specific in cleaving peptide bonds at the carboxy side of ...
UOM: 1 * 2 items
Catalog Number: (786-13)
Supplier: G-Biosciences
Description: SG-Chymotrypsinâ„¢ is a serine endopeptidase, which predominantly cleaves peptide bonds on the carboxy side of tyrosine, phenylalanine and tryptophan. I...
UOM: 1 * 2 items
Catalog Number: (786-11)
Supplier: G-Biosciences
Description: SG-Arginine-Câ„¢ endopeptidase (Clostripain, from C. histolyticum) specifically hydrolyses the carboxy peptide bond of arginine. SG-Arginine-Câ„¢ h...
UOM: 1 * 2 items
Catalog Number: (786-15)
Supplier: G-Biosciences
Description: SG-Glutamic-Câ„¢ is a serine endopeptidase, from S. aureus V8, that is highly specific for the cleavage of peptide bonds at the carboxy side of e...
UOM: 1 * 2 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 6 of 6
no targeter for Bottom