VWR® Magnetic Stirrers, Boxed Sets
Supplier: VWR
VWR®
442-0515EA 144 EUR
442-0515 442-0516 442-0517 442-0518 442-0522 442-0513 442-0514 442-0520 442-0521 442-0519
VWR® Magnetic Stirrers, Boxed Sets
Stirrers Magnetic Stirring Bars
Set Octahedral 1, PTFE covered, 12 octagonal magnetic stirring rods with centre ring, in practical, transparent box. Set includes two stirrers with each of following dimensions, L×Ø (mm): 13×8, 15×8, 25×10, 38×10, 51×10 and 64×10.
Set Cross-Shaped, PTFE covered, five cross-shaped magnetic stirring rods in practical, transparent box. Set includes one stirrer with each of following dimensions, L×Ø (mm): 10×10, 20×20, 25×25, 30×30 and 38×38.
Set Oval 1, PTFE covered, 12 oval magnetic stirring rods in practical, transparent box. Set includes two stirrers with each of following dimensions, L×Ø (mm): 10×5, 15×6, 25×10, 30×10 and 35×13. Set also includes one stirrer with each of following dimensions, L×Ø (mm): 50×17 and 70×27.
Set Oval 2, PTFE covered, 13 oval magnetic stirring rods in practical, transparent box. Set includes two stirrers with each of following dimensions, L×Ø (mm): 20×10, 25×12, 30×16, 35×16, 40×20 and 50×20. Set also includes one stirrer with following dimensions, L×Ø (mm): 70×20.
Set Coloured Micro, PTFE covered, 12 coloured micro magnetic stirring rods in practical, transparent box. Set includes one stirrer in each colour (blue, red, yellow) with following dimensions L×Ø (mm): 8×1,5, 5×2, 7×2 and 10×3.
Set Coloured Standard, PTFE covered, 24 coloured octagonal magnetic stirring rods with centre ring, in practical, transparent box. Set includes one stirrer in each colour (blue, red, yellow) with following dimensions L×Ø (mm): 13×3, 13×8, 15×8, 22×8, 25×8, 38×8, 51×8 and 75×13.
Set Polygon 1, PTFE covered, 18 cylindrical magnetic stirring bars in practical, transparent box. Set includes two stirrers with each of following dimensions, L×Ø (mm): 10×6, 15×6, 20×7, 25×8, 30×8, 40×8, 50×8, 60×10 and 80×10.
Set Polygon 2, PTFE covered, 18 cylindrical magnetic stirring bars in practical, transparent box. Set includes two stirrers with each of following dimensions, L×Ø (mm): 10×6, 15×4,5, 20×6, 25×6, 30×6, 40×8, 50×8, 60×10 and 70×10.
Set Mix 1, PTFE covered, 22 cylindrical magnetic stirring bars, 16 with removable pivot ring, in practical, transparent box. Set includes two stirrers with each of following dimensions, L×Ø (mm): 13×8, 13×10, 20×8, 25×8, 25×10, 38×8, 42×10, 51×8 and 51×10. Set also includes one stirrer with each of following dimensions, L×Ø (mm): 32×8, 32×10, 38×10 and 45×8.
Set Mix 2, PTFE covered. Set of 14 magnetic stirring bars: 4 micro and 10 octagonal with centre ring, in practical, transparent box. Set includes two stirrers with each of following dimensions, L×Ø (mm): 7×2, 10×3, 13×8,15×8, 25×10, 38×10 and 51×10.