You Searched For: Temperature Probes
85  results were found
SearchResultCount:"85"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (462-0553)
Supplier: VWR Collection
Description: External PT100 probe is used for with chillers and coolers, 6000 series chiller, non-refrigerated coolers, 4100 liquid-to-liquid coolers.
UOM: 1 * 1 items
Supplier: VWR Collection
Description: Temperature probe, VWR®
Supplier: VWR Collection
Description: Ultra-accurate K-type probe, Surface, Connectivity: SMP, −65…+400 °C, Ø×L: 1,5×100 mm
Supplier: VWR Collection
Description: External PT100 probes is for monitoring the temperature of an external application.
Supplier: VWR Collection
Description: Ultra accurate probe, VWR®, Air, −50…+400 °C, Ø×L: 3×150 mm
Supplier: VWR Collection
Description: Temperature probe, VWR®, Liquids, air, gas & semi-solids, −50…+700 °C, Ø×L: 3,30×190,5 mm
Supplier: VWR Collection
Description: Remote temperature probes for circulator use with a chiller. Allows points of control to be switched from inside a chiller to a remote location. Compe...
Catalog Number: (620-2810)
Supplier: VWR Collection
Description: Optimise performance of thermometer.
UOM: 1 * 1 items
Catalog Number: (620-1754)
Supplier: VWR Collection
Description: Replacement probe, VWR®
UOM: 1 * 1 items
Catalog Number: (620-1666)
Supplier: VWR Collection
Description: Thermometer probe, VWR®, −50…+200 °C, 125 mm
UOM: 1 * 1 items
Catalog Number: (620-1987)
Supplier: VWR Collection
Description: Thermometer probe, Traceable®, Liquids, air, gas & semi-solids, −50...+150 °C, Ø×L: — mm
UOM: 1 * 1 items
Catalog Number: (NABE801200000)
Supplier: NABERTHERM GMBH
Description: This thermocouple is suitable for muffle furnaces.
UOM: 1 * 1 items
Supplier: Testo
Description: Accurate probe, Air, −50...+125 °C, Ø×L: 5×115 mm
Supplier: Testo
Description: Probe for infrared thermometer, −50...+250 °C, 680 mm, Ø×L: 25×680 mm
Supplier: Testo
Description: Precision probe, T, Surface, Connectivity: Thermocouple, −50…+350 °C, Ø×L: 5×112 mm
Supplier: Testo
Description: Temperature probe, −50...+150 °C, 125 mm, Ø×L: 4×125 mm
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 85
no targeter for Bottom