Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (12)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
digital-thermometers-for-special-applications
SearchPresentationType-VERTICAL
SearchResultCount:"20"
  List View Grid View Easy View (new)
Sort by:
Supplier:  VWR Collection
Description:   These digital thermometers are designed for monitoring temperatures in refrigerators, freezers, water baths, heating blocks and incubators. Bottle pro...
Supplier:  VWR Collection
Description:   This high accuracy thermometer monitors both ambient air and the temperature inside a refrigerator or freezer. It reads to 0,01° while monitoring MIN/...
UOM:  1 * 1 items
Supplier:  VWR Collection
Description:   These highly accurate thermometers monitor temperatures simultaneously in refrigerators and freezers, two refrigerator locations. Temperature-buffered...
Supplier:  VWR Collection
Description:   Memory card reader (supplied) plugs into any USB port for an instant report. Text report includes date/time of day and sensor readings.
Catalog Number: (620-9100)
Supplier:  VWR Collection
Description:   This thermometer ensures proper cleaning by providing proof that the temperature regulation of achieving 82 °C is met during dishwashing. Designed for...
UOM:  1 * 1 items
Supplier:  VWR Collection
Description:   These thermometers accurately monitor temperatures in freezers, refrigerators, incubators and environmental chambers. Bottle and solution insulate the...
UOM:  1 * 1 items
Supplier:  VWR Collection
Description:   This radio-signal thermometer can read refrigerator temperatures from up to 100 feet away. They can provide readings from up to three remote refrigera...
UOM:  1 * 1 items
Supplier:  VWR Collection
Description:   These USB Traceable® excursion trac LN2 data logging thermometers monitor the temperature in the liquid nitrogen Dewars, covering cryogenic...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 20
  1  2  3  Next