You Searched For: Digital Thermometers
161  results were found
SearchResultCount:"161"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (620-2809)
Supplier: VWR Collection
Description: Simultaneously displays temperature, timer, and current alarms.
UOM: 1 * 1 items
Supplier: VWR Collection
Description: This waterproof and shockproof thermometer is a safe replacement for any hazardous mercury instrument. The probe is resistant to most chemicals.
Catalog Number: (620-2102)
Supplier: VWR Collection
Description: This thermometer captures temperature readings of any liquid, solid and semi-solid surfaces in less than a second. These non invasive, 'no touch' temp...
UOM: 1 * 1 items
Catalog Number: (620-1826)
Supplier: VWR Collection
Description: These thermometers accurately monitor temperatures in freezers, refrigerators, incubators and environmental chambers. Bottle and solution insulate the...
UOM: 1 * 1 items
Catalog Number: (620-1806)
Supplier: VWR Collection
Description: This HACCP - Hazard Analysis and Critical Control Point - thermometer is designed for food laboratories, food processing, food preparation and brewing...
UOM: 1 * 1 items
Supplier: VWR Collection
Description: Digital thermometers are designed to accurately monitor the temperature of fridges, freezers, incubators, water baths or during transport, in one or t...
Supplier: VWR Collection
Description: These highly accurate logging thermometers are designed to monitor temperatures in refrigerators, freezers, water baths, heating blocks and incubators...
Supplier: VWR Collection
Description: Accurate measurements for laboratories, quality management, and monitoring of production processes. Application: Food (HACCP), medical/pharmaceutical ...
Supplier: VWR Collection
Description: Data logging thermometers with user-defined timing intervals. Ideal for monitoring ultra-low temperature freezers.
Sale
Supplier: VWR Collection
Description: These high accuracy digital thermometers, with chemical resistant probes, monitor the temperature of pharmaceuticals, vaccines, chemicals, blood and b...
Catalog Number: (620-1975)
Supplier: VWR Collection
Description: Digital thermometer with extra long waterproof stainless steel 500 mm probe and 2,5 metre cable.
UOM: 1 * 1 items
Catalog Number: (620-2134)
Supplier: VWR Collection
Description: Digital stem thermometer suitable for field studies, wet lab applications, wash-down areas, food testing and tanks.
UOM: 1 * 1 items
Catalog Number: (620-2675)
Supplier: VWR Collection
Description: Precise universal thermometer, with BNC connection, without sensor. Pure measurement with a focus on precision, speed and reliability packaged in a co...
UOM: 1 * 1 items
Supplier: VWR Collection
Description: These highly accurate thermometers monitor temperatures simultaneously in refrigerators and freezers, two refrigerator locations. Temperature-buffered...
Catalog Number: (620-2094)
Supplier: VWR Collection
Description: This waterproof thermometer captures 10 temperature readings, stores them in memory, and recalls them on command. For food, agricultural, petrochemica...
UOM: 1 * 1 items
Catalog Number: (620-2813)
Supplier: VWR Collection
Description: Surface probe stem thermometer is ideal for cooking, laboratories and hot plates.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 161
no targeter for Bottom