Storage tubes, low volume, 2D barcoded, Matrix®
Supplier: Thermo Fisher Scientific
Matrix®
479-1193EA 341 EUR
479-1193 479-2045
Storage tubes, low volume, 2D barcoded, Matrix®
Vials
Virgin Class VI medical grade PP. For storage in virtually any laboratory condition, from room temperature through the vapour phase of liquid nitrogen. A permanently bonded, unique 2D barcode is laser-etched onto the base of every tube for secure identification and sample tracking.
  • All barcodes are checked against the database of previously assigned codes to prevent duplicates
  • Every tube is leak tested
  • Internally threaded single-piece caps with moulded-in gaskets that cannot be over-tightened manually or with automated capping tools
  • Free from DNA, RNAse, DNAse and endotoxins
  • Temperature resistant from −180 to +121 °C
For storage in virtually any laboratory condition, from room temperature through the vapour phase of liquid nitrogen. A permanently bonded, unique 2D barcode is laser-etched onto the base of every tube for secure identification and sample tracking.
  • All barcodes are checked against the database of previously assigned codes to prevent duplicates
  • Every tube is leak tested
  • Internally threaded single-piece caps with moulded-in gaskets that cannot be over-tightened manually or with automated capping tools
  • Free from DNA, RNAse, DNAse and endotoxins
  • Temperature resistant from -180 to +121 °C