Protein MW markers, mid/low range, Ultra Pure Grade
J450-200ULEA 151 EUR
VWRCJ450-200UL
Protein MW markers, mid/low range, Ultra Pure Grade
Mid/low range protein molecular weight marker offers seven bands ranging from 14,4 to 97,4 kDa. The 200 μl size provides sufficient material for 40 assays.
  • 7 bands ranging from 14,4 to 97,4 kDa
  • Ideal for polyacrylamide gel concentrations from 8 to 15% (37,5:1 acryl/bis)