Order Entry
Mini desiccator cabinets, SICCO, Protect
Mini desiccator cabinets, SICCO, Protect
Supplier:  Bohlender
Certificates
undefined
Mini desiccator cabinets, SICCO, Protect
Supplier:  Bohlender
Product Information
  • Catalog #
    Supplier #
    W×D×H ext.
    W×D×H int.
    Weight
    Pk
    Availability
    Price
  • Mini desiccator, Protect Premium (6,2 l)
    Mini desiccator, Protect Premium (6,2 l)
    467-0128
    V1942-06
    221×214×183 mm
    212×180×162 mm
    0,9 kg
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 209,00 €
    209,00 €
    New Customer Promotional Price
    1 * 1 items
       209,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Mini desiccator, Protect Basic (6,2 l)
    Mini desiccator, Protect Basic (6,2 l)
    467-0125
    V1842-06
    221×214×183 mm
    212×180×162 mm
    0,9 kg
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 186,00 €
    186,00 €
    New Customer Promotional Price
    1 * 1 items
       186,00 €
    Specifications:
    Specifications
    Environmentally Preferable