Rotating mixers, RM 5
Supplier: GLASWARENFABRIK KARL HECHT
Certificates
About this item
Specifications
- Catalog No:
- HECH40348026
- 445-2510
- HECH40349018
- Model:
- RM 5
- RM 5
- RM 5
- Shaking motion:
- Rotating
- Rotating
- Rotating
- Speed:
- 36 min⁻¹ fixed
- 36 min⁻¹ fixed
- 2 - 30 min⁻¹
- W×D×H:
- 332×180×95 mm
- 430×180×95 mm
- 436×358×105 mm
- No. of rollers:
- 5
- 5
- 10
- Weight:
- 3,2 kg
- 3,7 kg
- 3,8 kg
- Max. load:
- 2,0 kg
- 2,0 kg
- 2,0 kg
Specifications
Product Information
- Catalog #Supplier #SeqPkAvailabilityPrice
- Rotating mixer, with shorter rolls (length 200 mm)Rotating mixer, with shorter rolls (length 200 mm)HECH40348026403480260111 * 1 items672,00 €Specifications:
More
Specifications
ModelRM 5Environmentally PreferableShaking motionRotatingSpeed36 min⁻¹ fixedW×D×H332×180×95 mmNo. of rollers5Weight3,2 kgMax. load2,0 kg - Rotating mixer, with 5 fixed synchronous turning eccentric PVC rolls (length 330 mm)Rotating mixer, with 5 fixed synchronous turning eccentric PVC rolls (length 330 mm)445-2510403480100211 * 1 items681,00 €Specifications:
More
Specifications
ModelRM 5Environmentally PreferableShaking motionRotatingSpeed36 min⁻¹ fixedW×D×H430×180×95 mmNo. of rollers5Weight3,7 kgMax. load2,0 kg - Rotating mixer, with 10 rollers (length 324 mm)Rotating mixer, with 10 rollers (length 324 mm)HECH40349018403490180311 * 1 items1.530,00 €Specifications:
More
Specifications
ModelRM 5Environmentally PreferableShaking motionRotatingSpeed2 - 30 min⁻¹W×D×H436×358×105 mmNo. of rollers10Weight3,8 kgMax. load2,0 kg