Order Entry
VWR®, Sealing Film for Automation
undefined
VWR®, Sealing Film for Automation
  •  
    Seq
    Optical property
    Temperature Range
    Thickness
    Sterile
    Pk
  • Sealing film, polyolefin, DNase- and RNase-free
    Sealing film, polyolefin, DNase- and RNase-free
    731-0308
    01
    Optically clear
    –80...+105 °C
    101,6 µm
    -
    1 Roll
    1 * 1 Roll
    1 * 1 Roll - 1 180,00 €
    1 180,00 €
    1 * 1 Roll
       1 180,00 €
    Specifications
    Environmentally Preferable
  • Sealing foil, aluminium
    Sealing foil, aluminium
    731-0307
    02
    Light blocking
    –80...+120 °C
    75,7 µm
    -
    1 Roll
    1 * 1 Roll
    1 * 1 Roll - 778,00 €
    778,00 €
    1 * 1 Roll
       778,00 €
    Specifications
    Environmentally Preferable
  • Sealing film, rayon
    Sealing film, rayon
    731-0306
    03
    Light blocking
    –20...+80 °C
    139,7 µm
    -
    1 Roll
    1 * 1 Roll
    1 * 1 Roll - 861,00 €
    861,00 €
    1 * 1 Roll
       861,00 €