Single channel pipettes, mechanical, variable volume, with RFID tag, Pipet-Lite™ XLS+™
Supplier: Mettler-Toledo
Pipet-Lite™
RAINSL-300XLSEA 0 EUR
RAINSL-300XLS RAIN17014409 RAIN17014408 RAIN17014407 RAIN17014405 RAINL-5000XLS RAIN17014393 RAIN17014392 RAIN17014391 RAIN17014390 RAIN17014414 RAIN17011783 RAIN17014413 RAIN17014412 RAIN17014411 RAINL-20MLXLS RAIN17014410 RAIN17011788 RAIN17011801 RAINL-10MLXLS RAINSL-5000XLS RAIN17014384 RAIN17014382 RAINSL-10MLXLS RAIN17011795 RAIN17011790 RAIN17014388
Single channel pipettes, mechanical, variable volume, with RFID tag, Pipet-Lite™ XLS+™
Pipettes Micropipettes
This ergonomic manual pipette with comfortable handle ensure light and smooth operation, and significantly reduce the risk of repetitive strain injuries.
  • Ergonomic handle design
  • Improved volume adjustment lock prevents drift
  • Built-in shock absorber inside the tip ejector reduces thumb impact
  • Available with LTS, and universal tip/shaft configuration for use with universal tips
  • Corrosion free, easy to clean tip ejector
  • RFID tags for asset management
  • New 'stictionless' sealing system
  • Autoclavable (shaft and tip ejector)
Pipet-Lite XLS is the world’s first pipette equipped with RFID tags for advanced calibration and asset management. Tip shaft options included low-force LTS for improved ergonomics and universal-fit.