Order Entry
Viton Transfer Pump Tubing, Viton®, 1/4" ID x 5/16" OD; 25 ft
  MFLX06434-04
CE certificate
undefined
Viton Transfer Pump Tubing, Viton®, 1/4" ID x 5/16" OD; 25 ft
  MFLX06434-04
CE certificate
121,00 €
  1 * 25 Feet
  • Clarity:
  • Opaque
  • Opaque
  • Opaque
  • Opaque
  • Opaque
  • Opaque
  • Opaque
  • Opaque
  • Opaque
  • GMP compliant:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Max. pressure:
  • 1,1721092 bar (17 psi)
  • 1,378952 bar (20 psi)
  • 1,2410568 bar (18 psi)
  • 1,3100044 bar (19 psi)
  • 1,034214 bar (15 psi)
  • 1,1031616 bar (16 psi)
  • 1,1721092 bar (17 psi)
  • 1,5857948 bar (23 psi)
  • 1,4478996 bar (21 psi)
  • Max. temperature:
  • 205 °C (400 °F)
  • 205 °C (400 °F)
  • 205 °C (400 °F)
  • 205 °C (400 °F)
  • 205 °C (400 °F)
  • 205 °C (400 °F)
  • 205 °C (400 °F)
  • 205 °C (400 °F)
  • 205 °C (400 °F)
  • Min. temperature:
  • –32 °C (–25 °F)
  • –32 °C (–25 °F)
  • –32 °C (–25 °F)
  • –32 °C (–25 °F)
  • –32 °C (–25 °F)
  • –32 °C (–25 °F)
  • –32 °C (–25 °F)
  • –32 °C (–25 °F)
  • –32 °C (–25 °F)
  • Sterilisation method autoclave:
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • Sterilisation method EtO:
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • Sterilisation method gamma:
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • Working pressure:
  • 17 psi
  • 20 psi
  • 18 psi
  • 19 psi
  • 15 psi
  • 16 psi
  • 17 psi
  • 23 psi
  • 21 psi
  •  
keep light sensitive samples safe.
  • Global market leader in range of formulations for most extensive chemical compatibility
Applications: Acid and solvent transfer; high-temperature work.
Characteristics: Opaque black.
Certification: Tubing is manufactured according to GMP.
Temperature range: –25 to 399 °F (–32 to 204 °C)
Sterilize: Not recommended.
  • Tubing ext.Ø:
    7,9 mm (0,3125")
  • Tubing int.Ø:
    6,35 mm (0,25")
  • Wall thickness:
    0,8 mm
  • Max. pressure:
    1,1721092 bar (17 psi)
  • Max. temperature:
    205 °C (400 °F)
  • Colour:
    Black
  • Description:
    Transfer Pump Tubing, Viton®, 1/4" ID x 5/16" OD; 25 ft
  • Length:
    7,6 m
  • Environmentally Preferable:
  • GMP compliant:
    Yes
  • Sterilisation method autoclave:
    No
  • Sterilisation method EtO:
    No
  • Sterilisation method gamma:
    No
  • Min. temperature:
    –32 °C (–25 °F)
  • Working pressure:
    17 psi
  • Clarity:
    Opaque
  • Tubing material:
    Viton®
  • Pk:
    25 Feet