NeutraKlean - new sterile ready to use detergent from Contec
Keep your mind at ease and your A & B cleanrooms clean with this new sterile detergent
  • Gentle on surfaces: Neutral pH, low residue formulation
  • Gentle on people and the environment: No CLP hazard classification
  • Easy to use: Easy tear bags, no further dilution required, available in 1 l spray bottle or 5 l canister
  • Formulated with purified water (Ph. Eur.)
  • Validated gamma irradiation, minimum 25 kGy
  • Shelf life of 2 years when unopened, with an in-use shelf life of 6 months for the 1 l bag-in-bottle
Description Pk Cat. No. Price Quantity
NeutraKlean sterile trigger spray 6x 1 l 141-1305E
NeutraKlean sterile canister 2x 5 l 141-1306E